Assay Details
Target Gene Details
Entrez Gene ID: | 8611 |
Gene Name: | phospholipid phosphatase 1 |
Gene Aliases: |
LLP1a, LPP1, PAP-2a, PAP2, PPAP2A |
Location: |
Chr.5:55424842-55535370 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP1 | NM_003711.3 | 6 | 1360 | NP_003702.2 |
| NM_176895.2 | 6 | 1363 | NP_795714.1 | |
| NR_103485.1 | 7 | 1515 | ||
| XM_006714724.2 | 5 | 1500 | XP_006714787.1 | |
| AB000888.1 | BAA22593.1 | |||
| AF014402.1 | 6 | 1252 | AAC16032.1 | |
| AF014403.1 | 6 | 1255 | AAC16033.1 | |
| AF017116.1 | AAC32041.1 | |||
| BC039847.1 | 6 | 1224 | AAH39847.1 | |
| BC117133.1 | ||||
| BC143281.1 | ||||
| BQ019387.1 | 1 | 326 |
Target Gene Details
Entrez Gene ID: | 23517 |
Gene Name: | Ski2 like RNA helicase 2 |
Gene Aliases: |
Dob1, KIAA0052, Mtr4, fSAP118 |
Location: |
Chr.5:55307748-55425581 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKIV2L2 | NM_015360.4 | 27 | 3773 | NP_056175.3 |
| BC065258.1 | 27 | 3512 | AAH65258.1 |
More Information
Set Membership: |
|

Genomic Map