Assay Details
Target Gene Details
Entrez Gene ID: | 122769 |
Gene Name: | leucine rich repeat protein 1 |
Gene Aliases: |
4-1BBLRR, LRR-1, PPIL5 |
Location: |
Chr.14:49598697-49614672 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRR1 | NM_152329.3 | 1 | 163 | NP_689542.2 |
| NM_203467.1 | 1 | 163 | NP_982292.1 | |
| NR_037792.1 | 1 | 163 | ||
| NR_037793.1 | 1 | 163 | ||
| BC030142.2 | 1 | 99 | AAH30142.1 | |
| BI825814.1 | 1 | 90 | ||
| BI826606.1 | 1 | 163 | ||
| BI829295.1 | 1 | 100 | ||
| BX648029.1 | 1 | 86 | CAH10374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map