Assay Details
Target Gene Details
Entrez Gene ID: | 6329 |
Gene Name: | sodium voltage-gated channel alpha subunit 4 |
Gene Aliases: |
CMS16, HOKPP2, HYKPP, HYPP, NAC1A, Na(V)1.4, Nav1.4, SkM1 |
Location: |
Chr.17:63938554-63972918 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN4A | NM_000334.4 | 24 | 7749 | NP_000325.4 |
| M81758.1 | 24 | 7749 | AAA60554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833511 | Chr.17:63865995 - 64064305 on Build GRCh38 | Loss |
|
| nsv517272 | Chr.17:63814307 - 64020132 on Build GRCh38 | Gain+Loss |
|
| esv3641021 | Chr.17:63933626 - 63991855 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map