Assay Details
Target Gene Details
Entrez Gene ID: | 259232 |
Gene Name: | sodium leak channel, non-selective |
Gene Aliases: |
CLIFAHDD, CanIon, IHPRF, IHPRF1, INNFD, VGCNL1, bA430M15.1 |
Location: |
Chr.13:101053774-101417206 on Build GRCh38 |
Assay Gene Location: | Within Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NALCN | NM_052867.2 | 44 | 6560 | NP_443099.1 |
| XM_011521067.2 | 44 | 6500 | XP_011519369.1 | |
| AF339780.1 | 1 | 432 | ||
| AK002089.1 | 3 | 1627 | ||
| AK055506.1 | 11 | 2617 | ||
| AK126621.1 | 25 | 4024 | ||
| AK126718.1 | 17 | 3161 | ||
| AK172752.1 | 7 | 2091 | BAD18738.1 |
Target Gene Details
Entrez Gene ID: | 100885778 |
Gene Name: | NALCN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:100708325-101059286 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NALCN-AS1 | NR_047687.1 | |||
| AK025806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1149 | Chr.13:101042454 - 101087249 on Build GRCh38 | Deletion |
|
| nsv832699 | Chr.13:100951779 - 101142880 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map