Assay Details
Target Gene Details
Entrez Gene ID: | 202309 |
Gene Name: | GRB2-binding adaptor protein, transmembrane |
Gene Aliases: |
C5orf29 |
Location: |
Chr.5:58491435-58496358 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAPT | NM_001304428.1 | 3 | 465 | NP_001291357.1 |
| NM_001304429.1 | 3 | 458 | NP_001291358.1 | |
| NM_001304431.1 | 3 | 366 | NP_001291360.1 | |
| NM_152687.3 | 3 | 303 | NP_689900.1 | |
| AK090960.1 | 3 | 237 | BAC03557.1 | |
| BC063534.1 | AAH63534.1 | |||
| DA008727.1 | 3 | 366 | ||
| DA347824.1 | 3 | 419 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4844 | Chr.5:58476094 - 58509159 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map