Assay Details
Target Gene Details
Entrez Gene ID: | 161142 |
Gene Name: | family with sequence similarity 71 member D |
Gene Aliases: |
C14orf54 |
Location: |
Chr.14:67188365-67228550 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71D | NM_173526.3 | 5 | 1012 | NP_775797.2 |
| XM_011536500.1 | 5 | 976 | XP_011534802.1 | |
| XM_011536502.1 | 5 | 911 | XP_011534804.1 | |
| XM_011536503.2 | 4 | 1953 | XP_011534805.1 | |
| AK093449.1 | 5 | 976 | BAC04169.1 | |
| BC050401.1 | 5 | 944 | AAH50401.1 |
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751285 | Chr.14:66898030 - 67420064 on Build GRCh38 | Gain |
|
| nsv832821 | Chr.14:67123701 - 67361452 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map