Assay Details
Target Gene Details
Entrez Gene ID: | 29072 |
Gene Name: | SET domain containing 2 |
Gene Aliases: |
HBP231, HIF-1, HIP-1, HSPC069, HYPB, KMT3A, LLS, SET2, p231HBP |
Location: |
Chr.3:47016408-47164109 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD2 | NM_014159.6 | NP_054878.5 | ||
| XM_011533631.2 | XP_011531933.1 | |||
| XM_011533632.2 | XP_011531934.1 | |||
| XM_011533633.2 | XP_011531935.1 | |||
| XM_011533634.1 | XP_011531936.1 | |||
| XM_017006270.1 | XP_016861759.1 | |||
| AB051519.2 | BAB21823.2 | |||
| AJ238403.1 | CAC28349.1 | |||
| AK026837.1 | ||||
| AK095494.1 | ||||
| AL713692.1 | CAD28492.1 | |||
| AL831959.1 | CAD38601.2 | |||
| AY576987.1 | AAT77612.1 | |||
| AY576988.1 | AAT77613.1 | |||
| BC090954.1 | AAH90954.1 | |||
| BC117162.1 | ||||
| BC117164.1 | ||||
| BX649110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007714 | Chr.3:47042897 - 47290080 on Build GRCh38 | Gain |
|
| nsv997261 | Chr.3:47013276 - 47092190 on Build GRCh38 | Gain |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
| esv3596073 | Chr.3:46936296 - 47131646 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map