Assay Details
Target Gene Details
Entrez Gene ID: | 2059 |
Gene Name: | epidermal growth factor receptor pathway substrate 8 |
Gene Aliases: |
DFNB102 |
Location: |
Chr.12:15620141-15789576 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS8 | NM_004447.5 | 17 | 2169 | NP_004438.3 |
| XM_011520605.2 | 18 | 2229 | XP_011518907.1 | |
| AK291777.1 | 17 | 1974 | ||
| AK292931.1 | 17 | 1826 | ||
| AK301834.1 | 10 | 1159 | ||
| AK304860.1 | 17 | 2169 | ||
| AK316134.1 | 10 | 1078 | ||
| AK316239.1 | 10 | 1351 | ||
| BC030010.2 | 17 | 1981 | AAH30010.1 | |
| U12535.1 | 17 | 1941 | AAA62280.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map