Assay Details
Target Gene Details
Entrez Gene ID: | 2752 |
Gene Name: | glutamate-ammonia ligase |
Gene Aliases: |
GLNS, GS, PIG43, PIG59 |
Location: |
Chr.1:182378093-182392206 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLUL | NM_001033044.3 | NP_001028216.1 | ||
| NM_001033056.3 | NP_001028228.1 | |||
| NM_002065.6 | NP_002056.2 | |||
| XM_006711278.1 | XP_006711341.1 | |||
| AK122784.1 | ||||
| AK301600.1 | ||||
| AL161952.1 | 2 | 2347 | CAB82304.1 | |
| AY486122.1 | AAS57904.1 | |||
| AY513283.1 | AAT08036.1 | |||
| AY947343.1 | ||||
| BC010037.2 | AAH10037.1 | |||
| BC011700.2 | AAH11700.1 | |||
| BC011852.2 | AAH11852.1 | |||
| BC018992.1 | AAH18992.1 | |||
| BC031964.1 | AAH31964.1 | |||
| BC051726.1 | AAH51726.1 | |||
| BC127883.1 | ||||
| BX537384.1 | CAD97626.1 | |||
| S70290.1 | AAB30693.1 | |||
| X59834.1 | CAA42495.1 | |||
| Y00387.2 | CAA68457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946532 | Chr.1:182384061 - 182385635 on Build GRCh38 | Duplication |
|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map