Assay Details
Target Gene Details
Entrez Gene ID: | 4694 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A1 |
Gene Aliases: |
CI-MWFE, MWFE, ZNF183 |
Location: |
Chr.X:119871771-119876666 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA1 | NM_004541.3 | 1 | 198 | NP_004532.1 |
| AB451304.1 | 1 | 57 | ||
| AB451435.1 | 1 | 57 | ||
| BC000266.1 | 1 | 95 | AAH00266.1 | |
| BQ680674.1 | 1 | 99 | ||
| CR456889.1 | 1 | 57 | CAG33170.1 | |
| U54993.1 | 1 | 90 | AAD00084.1 | |
| X81900.1 | 1 | 74 | CAA57489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7057 | Chr.X:119862293 - 119894925 on Build GRCh38 | Insertion |
|
| esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map