Assay Details
Target Gene Details
Entrez Gene ID: | 55922 |
Gene Name: | NFKB repressing factor |
Gene Aliases: |
ITBA4, NRF |
Location: |
Chr.X:119588337-119606464 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKRF | NM_001173487.1 | 4 | 3097 | NP_001166958.1 |
| NM_001173488.1 | 2 | 3483 | NP_001166959.1 | |
| NM_017544.3 | 3 | 2999 | NP_060014.2 | |
| XM_011531365.2 | 4 | 3629 | XP_011529667.1 | |
| AA845622.1 | 1 | 265 | ||
| AJ011812.2 | 2 | 3483 | CAB56459.1 | |
| AY208891.1 | 3 | 2999 | AAP43025.1 | |
| BC047878.2 | 3 | 2966 | AAH47878.1 | |
| EF165092.1 | 1 | 2557 | ||
| EF216689.1 | 2 | 3483 | ||
| Y07707.1 | 1 | 1474 | CAA68976.1 |
More Information
Set Membership: |
|

Genomic Map