Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Gene Details
Entrez Gene ID: | 649446 |
Gene Name: | DLGAP1 antisense RNA 1 |
Gene Aliases: |
HsT914 |
Location: |
Chr.18:3594114-3597379 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP1-AS1 | NR_024101.1 | 3 | 858 | |
| AI275162.1 | 1 | 166 | ||
| AK093095.1 | 3 | 858 | BAC04052.1 | |
| BX647607.1 | 1 | 2798 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv184n21 | Chr.18:3526777 - 5930980 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map