Assay Details
Target Gene Details
Entrez Gene ID: | 959 |
Gene Name: | CD40 ligand |
Gene Aliases: |
CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L |
Location: |
Chr.X:136648177-136660390 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD40LG | NM_000074.2 | 5 | 1645 | NP_000065.1 |
| BC071754.1 | 5 | 1645 | AAH71754.1 | |
| L07414.1 | 5 | 1612 | AAA35662.1 | |
| X67878.1 | 5 | 1618 | CAA48077.1 | |
| X68550.1 | 5 | 1629 | CAA48554.1 |
More Information
Set Membership: |
|

Genomic Map