Assay Details
Target Gene Details
Entrez Gene ID: | 27336 |
Gene Name: | HIV-1 Tat specific factor 1 |
Gene Aliases: |
TAT-SF1, TATSF1, dJ196E23.2 |
Location: |
Chr.X:136496932-136512346 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTATSF1 | NM_001163280.1 | NP_001156752.1 | ||
| NM_014500.4 | NP_055315.2 | |||
| XM_005262404.3 | XP_005262461.1 | |||
| AB209303.1 | BAD92540.1 | |||
| AK225870.1 | ||||
| AK299403.1 | ||||
| BC009896.2 | AAH09896.1 | |||
| BT006886.1 | AAP35532.1 | |||
| U76992.1 | AAB18823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577408 | Chr.X:136488033 - 136507698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map