Assay Details
Target Gene Details
Entrez Gene ID: | 81617 |
Gene Name: | calcium binding protein 39 like |
Gene Aliases: |
MO25-BETA, MO2L, bA103J18.3 |
Location: |
Chr.13:49308628-49444346 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAB39L | NM_001079670.2 | 11 | 1789 | NP_001073138.1 |
| NM_001287337.1 | 12 | 1927 | NP_001274266.1 | |
| NM_001287338.1 | 12 | 1922 | NP_001274267.1 | |
| NM_001287339.1 | 10 | 1591 | NP_001274268.1 | |
| NM_030925.3 | 9 | 1840 | NP_112187.2 | |
| XM_011535254.2 | 13 | 2331 | XP_011533556.1 | |
| XM_011535255.2 | 11 | 1811 | XP_011533557.1 | |
| XM_011535256.2 | 12 | 2076 | XP_011533558.1 | |
| XM_017020785.1 | 12 | 2203 | XP_016876274.1 | |
| XM_017020786.1 | 13 | 2340 | XP_016876275.1 | |
| XM_017020787.1 | 13 | 2593 | XP_016876276.1 | |
| XM_017020788.1 | 14 | 2469 | XP_016876277.1 | |
| AK022639.1 | 8 | 1385 | BAB14147.1 | |
| AK309011.1 | 6 | 1115 | ||
| BX647518.1 | 11 | 1726 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3449183 | Chr.13:49309415 - 49310713 on Build GRCh38 | Duplication |
|
| nsv832607 | Chr.13:49182322 - 49346758 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map