Assay Details
Target Gene Details
Entrez Gene ID: | 2954 |
Gene Name: | glutathione S-transferase zeta 1 |
Gene Aliases: |
GSTZ1-1, MAAI, MAI |
Location: |
Chr.14:77320887-77331597 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTZ1 | NM_001312660.1 | 10 | 1190 | NP_001299589.1 |
| NM_145870.2 | 9 | 1004 | NP_665877.1 | |
| NM_145871.2 | 8 | 878 | NP_665878.2 | |
| XM_011536670.2 | 9 | 1605 | XP_011534972.1 | |
| XM_011536671.2 | 8 | 788 | XP_011534973.1 | |
| XM_017021243.1 | 8 | 1275 | XP_016876732.1 | |
| AB209360.1 | 6 | 5130 | BAD92597.1 | |
| AJ001838.1 | 9 | 825 | CAA05045.1 | |
| AK094344.1 | 2 | 2118 | ||
| BC001453.1 | 9 | 880 | AAH01453.1 | |
| BG330365.1 | ||||
| U86529.1 | 9 | 825 | AAB96392.1 |
More Information
Set Membership: |
|

Genomic Map