Assay Details
Target Gene Details
Entrez Gene ID: | 4355 |
Gene Name: | membrane palmitoylated protein 2 |
Gene Aliases: |
DLG2 |
Location: |
Chr.17:43875357-43909711 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPP2 | NM_001278370.1 | 13 | 3871 | NP_001265299.1 |
| NM_001278371.1 | 12 | 3807 | NP_001265300.1 | |
| NM_001278372.1 | 14 | 4242 | NP_001265301.1 | |
| NM_001278373.1 | 12 | 4106 | NP_001265302.1 | |
| NM_001278374.1 | 12 | 4051 | NP_001265303.1 | |
| NM_001278375.1 | 12 | 3916 | NP_001265304.1 | |
| NM_001278376.2 | 12 | 3800 | NP_001265305.2 | |
| NM_001278381.1 | 14 | 4503 | NP_001265310.1 | |
| NM_005374.4 | 13 | 4170 | NP_005365.4 | |
| XM_011524827.2 | 14 | 3965 | XP_011523129.2 | |
| AL136554.1 | 13 | 3750 | CAB66489.1 | |
| BC030287.1 | 13 | 3973 | AAH30287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640640 | Chr.17:43868102 - 43882275 on Build GRCh38 | Gain |
|
| esv3640639 | Chr.17:43862438 - 43888388 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73985803,rs77504473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map