Assay Details
Target Gene Details
Entrez Gene ID: | 80709 |
Gene Name: | AT-hook transcription factor |
Gene Aliases: |
- |
Location: |
Chr.9:114334149-114398498 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKNA | NM_001317950.1 | 22 | 4810 | NP_001304879.1 |
| NM_001317952.1 | 21 | 4368 | NP_001304881.1 | |
| NM_030767.5 | 22 | 4755 | NP_110394.3 | |
| XM_005252244.2 | 24 | 4927 | XP_005252301.1 | |
| XM_005252245.1 | 23 | 4785 | XP_005252302.1 | |
| XM_005252247.4 | 22 | 4806 | XP_005252304.1 | |
| XM_005252248.2 | 21 | 4827 | XP_005252305.1 | |
| XM_006717294.1 | 22 | 4739 | XP_006717357.1 | |
| XM_011519063.2 | 21 | 4418 | XP_011517365.1 | |
| XM_011519065.2 | 22 | 4398 | XP_011517367.2 | |
| XM_017015172.1 | 23 | 4525 | XP_016870661.1 | |
| AK024431.1 | 20 | 3703 | BAB15721.1 | |
| AK074040.1 | 21 | 3798 | BAB84866.1 | |
| AK131082.2 | 22 | 4811 | BAC85132.1 | |
| AK160382.1 | 22 | 4813 | BAD18725.1 | |
| AL713755.1 | 5 | 3244 | ||
| AL833220.1 | 19 | 3232 | ||
| AY703039.1 | 23 | 4410 | AAU34186.1 | |
| AY703040.1 | 21 | 4368 | AAU34187.1 | |
| AY703043.1 | 21 | 4728 | AAU34190.1 | |
| AY703044.1 | 20 | 4422 | AAU34191.1 | |
| AY703045.1 | 13 | 2501 | AAU34192.1 | |
| AY703046.1 | 9 | 1627 | AAU34193.1 | |
| BC042202.1 | 8 | 1485 | ||
| BC055285.1 | 22 | 4755 | AAH55285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv948n27 | Chr.9:114193988 - 114693895 on Build GRCh38 | Gain |
|
| esv2752277 | Chr.9:114323991 - 114338965 on Build GRCh38 | Gain |
|
| esv2761285 | Chr.9:114049617 - 114340988 on Build GRCh38 | Gain |
|
| nsv509319 | Chr.9:114297186 - 114380605 on Build GRCh38 | Insertion |
|
| dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
| nsv471320 | Chr.9:114284841 - 114346385 on Build GRCh38 | Gain |
|
| nsv615233 | Chr.9:114167820 - 114486199 on Build GRCh38 | Gain |
|
| dgv949n27 | Chr.9:114283521 - 114343856 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map