Assay Details
Target Gene Details
Entrez Gene ID: | 10160 |
Gene Name: | FERM, ARH/RhoGEF and pleckstrin domain protein 1 |
Gene Aliases: |
CDEP, FARP1-IT1, PLEKHC2, PPP1R75 |
Location: |
Chr.13:98142562-98449773 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARP1 | NM_001001715.3 | NP_001001715.2 | ||
| NM_001286839.1 | NP_001273768.1 | |||
| NM_005766.3 | NP_005757.1 | |||
| XM_011521046.2 | XP_011519348.1 | |||
| XM_017020312.1 | XP_016875801.1 | |||
| AB008430.1 | BAA24267.1 | |||
| AI273567.1 | ||||
| BC041595.1 | AAH41595.1 | |||
| BC071592.1 | AAH71592.1 | |||
| BM763139.1 | ||||
| CF994038.1 | ||||
| DB498534.1 |
Target Gene Details
Entrez Gene ID: | 140432 |
Gene Name: | ring finger protein 113B |
Gene Aliases: |
RNF161, ZNF183L1, bA10G5.1 |
Location: |
Chr.13:98175785-98177267 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF113B | NM_178861.4 | 1 | 414 | NP_849192.1 |
| AF539427.1 | 1 | 395 | AAN33063.1 | |
| BC017585.2 | 1 | 414 | AAH17585.1 | |
| BC025388.1 | 1 | 407 | AAH25388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045047 | Chr.13:98175009 - 98306412 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map