Assay Details
Target Gene Details
Entrez Gene ID: | 5407 |
Gene Name: | pancreatic lipase related protein 1 |
Gene Aliases: |
PLRP1 |
Location: |
Chr.10:116590415-116609175 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNLIPRP1 | NM_001303135.1 | NP_001290064.1 | ||
| NM_006229.3 | NP_006220.1 | |||
| XM_011539868.1 | XP_011538170.1 | |||
| XM_011539869.1 | 5 | 781 | XP_011538171.1 | |
| AK130236.1 | ||||
| AK307063.1 | 5 | 781 | ||
| BC025784.2 | AAH25784.1 | |||
| BP323340.1 | ||||
| CR749299.1 | 5 | 790 | CAH18154.1 | |
| CR749524.1 | 5 | 790 | CAH18337.1 | |
| CR749596.1 | 5 | 795 | CAH18394.1 | |
| M93283.1 | AAA59532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624660 | Chr.10:116578660 - 116596498 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map