Assay Details
Target Gene Details
| Entrez Gene ID: | 3067 | 
| Gene Name: | histidine decarboxylase | 
| Gene Aliases: | - | 
| Location: | Chr.15:50241945-50266049 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 15 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| HDC | NM_001306146.1 | 11 | 1694 | NP_001293075.1 | 
| NM_002112.3 | 12 | 1793 | NP_002103.2 | |
| XM_017022094.1 | 12 | 1662 | XP_016877583.1 | |
| XM_017022095.1 | 11 | 1563 | XP_016877584.1 | |
| XM_017022096.1 | 11 | 1447 | XP_016877585.1 | |
| XM_017022097.1 | 10 | 1745 | XP_016877586.1 | |
| XM_017022098.1 | 9 | 1265 | XP_016877587.1 | |
| AK027221.1 | 5 | 1196 | ||
| BC130527.1 | 12 | 1494 | ||
| BC144173.1 | 11 | 1395 | ||
| M60445.1 | 12 | 1502 | AAC41698.1 | |
| X54297.1 | 12 | 1553 | CAA38196.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1531 | Chr.15:50225808 - 50257078 on Build GRCh38 | Insertion |  SLC27A2  HDC | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top