Assay Details
Target Gene Details
Entrez Gene ID: | 7456 |
Gene Name: | WAS/WASL interacting protein family member 1 |
Gene Aliases: |
PRPL-2, WAS2, WASPIP, WIP |
Location: |
Chr.2:174559574-174682899 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WIPF1 | NM_001077269.1 | 5 | 860 | NP_001070737.1 |
| NM_003387.4 | 5 | 796 | NP_003378.3 | |
| XM_011511780.2 | 5 | 982 | XP_011510082.1 | |
| AF031588.1 | 5 | 804 | AAC03767.1 | |
| AF106062.1 | 1 | 125 | AAD45972.1 | |
| AK301271.1 | 5 | 801 | ||
| AK304194.1 | 5 | 813 | ||
| AL529597.3 | 5 | 828 | ||
| BC002914.2 | AAH02914.1 | |||
| BC110288.1 | 5 | 832 | AAI10289.1 | |
| BQ441937.1 | 3 | 597 | ||
| BX640870.1 | 5 | 860 | CAE45928.1 | |
| X86019.1 | 5 | 900 | CAA60014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3041 | Chr.2:174556013 - 174576388 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map