Assay Details
Target Gene Details
Entrez Gene ID: | 8697 |
Gene Name: | cell division cycle 23 |
Gene Aliases: |
ANAPC8, APC8, CUT23 |
Location: |
Chr.5:138187648-138213343 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC23 | NM_004661.3 | 16 | 1730 | NP_004652.2 |
| AB011472.1 | 16 | 1717 | BAA75628.1 | |
| AF053977.1 | 16 | 1697 | AAC70920.1 | |
| AF191341.1 | 16 | 1681 | AAF05755.1 | |
| AK223250.1 | 16 | 1718 | BAD96970.1 | |
| AK291610.1 | 16 | 1712 | ||
| AK304635.1 | 15 | 1594 | ||
| BC017713.1 | 16 | 1689 | AAH17713.1 | |
| BT009810.1 | 16 | 1681 | AAP88812.1 |
More Information
Set Membership: |
|

Genomic Map