Assay Details
Target Gene Details
Entrez Gene ID: | 4088 |
Gene Name: | SMAD family member 3 |
Gene Aliases: |
HSPC193, HsT17436, JV15-2, LDS1C, LDS3, MADH3 |
Location: |
Chr.15:67065698-67195195 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD3 | NM_005902.3 | 1 | 372 | NP_005893.1 |
| XM_011521559.2 | 1 | 531 | XP_011519861.1 | |
| AB209616.1 | BAD92853.1 | |||
| AI762628.1 | 1 | 372 | ||
| AK290881.1 | 2 | 297 | ||
| BC050743.1 | 1 | 290 | AAH50743.1 | |
| BF058230.1 | 1 | 372 | ||
| U68019.1 | 1 | 140 | AAB80960.1 | |
| U76622.1 | 1 | 111 | AAB18967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569796 | Chr.15:67030906 - 67073284 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map