Assay Details
Target Gene Details
Entrez Gene ID: | 57560 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
ATD2, SRTD2, WDR56 |
Location: |
Chr.3:160256986-160399532 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT80 | NM_001190241.1 | 21 | 4026 | NP_001177170.1 |
| NM_001190242.1 | 19 | 3660 | NP_001177171.1 | |
| NM_020800.2 | 20 | 3806 | NP_065851.1 | |
| AB037795.1 | 19 | 3550 | ||
| AL133045.1 | 5 | 1645 | CAB61372.1 | |
| CA421514.1 | 1 | 517 |
More Information
Set Membership: |
|

Genomic Map