Assay Details
Target Gene Details
Entrez Gene ID: | 480 |
Gene Name: | ATPase Na+/K+ transporting subunit alpha 4 |
Gene Aliases: |
ATP1A1, ATP1AL2 |
Location: |
Chr.1:160151562-160186977 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1A4 | NM_144699.3 | 1 | 237 | NP_653300.2 |
| AF506797.1 | 1 | 229 | AAQ07964.1 | |
| AK308286.1 | 1 | 195 | ||
| AL833335.1 | 1 | 210 | ||
| BC094801.1 | 1 | 188 | AAH94801.1 | |
| BX647234.1 | 1 | 211 | ||
| DB094047.1 | 1 | 237 |
More Information
Set Membership: |
|

Genomic Map