Assay Details
Target Gene Details
Entrez Gene ID: | 89910 |
Gene Name: | ubiquitin protein ligase E3B |
Gene Aliases: |
BPIDS, KOS |
Location: |
Chr.12:109477410-109543628 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3B | NM_130466.3 | 10 | 1379 | NP_569733.2 |
| NM_183415.2 | 10 | 1034 | NP_904324.1 | |
| XM_005253987.2 | 10 | 1259 | XP_005254044.1 | |
| XM_006719681.3 | 10 | 1592 | XP_006719744.1 | |
| XM_006719682.2 | 10 | 1592 | XP_006719745.1 | |
| XM_011538959.2 | 9 | 1141 | XP_011537261.1 | |
| XM_011538961.1 | 10 | 1592 | XP_011537263.1 | |
| XM_017020195.1 | 10 | 1802 | XP_016875684.1 | |
| AF130072.1 | 8 | 771 | ||
| AF251046.2 | 10 | 1349 | AAK28419.2 | |
| AK307385.1 | 9 | 913 | ||
| AL096740.1 | 10 | 1348 | CAH56410.1 | |
| BC032301.1 | 10 | 1012 | ||
| BC141880.1 | 10 | 1257 | ||
| BC150637.1 | 10 | 1010 | ||
| BX537403.1 | 10 | 1024 | CAD97645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630719 | Chr.12:109485009 - 109700854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map