Assay Details
Target Gene Details
| Entrez Gene ID: | 57129 | 
| Gene Name: | mitochondrial ribosomal protein L47 | 
| Gene Aliases: | CGI-204, L47mt, MRP-L47, NCM1 | 
| Location: | Chr.3:179588465-179604646 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 6 - Exon 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MRPL47 | NM_020409.2 | NP_065142.2 | ||
| NM_177988.1 | NP_817125.1 | |||
| AY212270.1 | AAO92749.1 | |||
| BC021575.1 | ||||
| BC032522.1 | AAH32522.1 | |||
| CA428787.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  non-DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top