Assay Details
Target Gene Details
Entrez Gene ID: | 28990 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
HT001 |
Location: |
Chr.3:131013873-131026854 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASTE1 | NM_001288950.1 | NP_001275879.1 | ||
NM_014065.3 | NP_054784.2 | |||
NR_110229.1 | ||||
XM_017006266.1 | XP_016861755.1 | |||
XM_017006267.1 | XP_016861756.1 | |||
XM_017006268.1 | 6 | 2245 | XP_016861757.1 | |
XM_017006269.1 | 6 | 2166 | XP_016861758.1 | |
AF113539.1 | AAF14876.1 | |||
AK027258.1 | ||||
AK027806.1 | BAB55383.1 | |||
AK294156.1 | ||||
AK301388.1 | ||||
BC030274.1 | ||||
BC103709.1 | AAI03710.1 | |||
BC110616.2 | AAI10617.1 |
Target Gene Details
Entrez Gene ID: | 27032 |
Gene Name: | ATPase secretory pathway Ca2+ transporting 1 |
Gene Aliases: |
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1 |
Location: |
Chr.3:130850500-131016712 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP2C1 | NM_001001485.2 | NP_001001485.1 | ||
NM_001001486.1 | NP_001001486.1 | |||
NM_001001487.1 | NP_001001487.1 | |||
NM_001199180.1 | NP_001186109.1 | |||
NM_001199182.1 | NP_001186111.1 | |||
NM_001199183.1 | NP_001186112.1 | |||
NM_001199185.1 | NP_001186114.1 | |||
XM_005247354.2 | XP_005247411.1 | |||
XM_005247355.2 | XP_005247412.1 | |||
XM_005247356.2 | XP_005247413.1 | |||
XM_005247358.2 | XP_005247415.1 | |||
XM_011512686.2 | XP_011510988.1 | |||
XM_017006164.1 | XP_016861653.1 | |||
AF181121.1 | AAF26296.1 | |||
AF225981.1 | AAF35375.1 | |||
AK074692.1 | BAC11142.1 | |||
AK296470.1 | ||||
AK299945.1 | ||||
AL833453.1 | 1 | 3186 | ||
AY268374.1 | AAP30008.1 | |||
AY268375.1 | AAP30009.1 | |||
BC028139.1 | AAH28139.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
