Assay Details
Target Gene Details
Entrez Gene ID: | 9512 |
Gene Name: | peptidase, mitochondrial processing beta subunit |
Gene Aliases: |
Beta-MPP, MPP11, MPPB, MPPP52, P-52 |
Location: |
Chr.7:103297412-103329902 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMPCB | NM_004279.2 | 8 | 936 | NP_004270.2 |
| XM_005250717.3 | 8 | 950 | XP_005250774.1 | |
| XM_006716181.3 | 8 | 949 | XP_006716244.1 | |
| AF054182.1 | 8 | 915 | AAC39915.1 | |
| AK000994.1 | 8 | 918 | ||
| AK057656.1 | 7 | 734 | ||
| AK098616.1 | 8 | 906 | ||
| AK289864.1 | 8 | 918 | ||
| AK297354.1 | 8 | 1301 | ||
| AL050216.1 | 4 | 385 | CAB43319.1 | |
| BC014079.2 | 8 | 876 | AAH14079.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831084 | Chr.7:103138461 - 103324523 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map