Assay Details
Target Gene Details
Entrez Gene ID: | 83959 |
Gene Name: | solute carrier family 4 member 11 |
Gene Aliases: |
BTR1, CDPD1, CHED, CHED2, NABC1, dJ794I6.2 |
Location: |
Chr.20:3227417-3241483 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A11 | NM_001174089.1 | 20 | 2712 | NP_001167560.1 |
| NM_001174090.1 | 20 | 2862 | NP_001167561.1 | |
| NM_032034.3 | 19 | 2704 | NP_114423.1 | |
| NR_135000.1 | 20 | 2706 | ||
| XM_005260856.4 | 19 | 3086 | XP_005260913.1 | |
| XM_005260857.1 | 20 | 2731 | XP_005260914.1 | |
| XM_011529383.2 | 20 | 4307 | XP_011527685.1 | |
| XM_011529384.1 | 20 | 2949 | XP_011527686.1 | |
| XM_011529385.1 | 20 | 2776 | XP_011527687.1 | |
| XM_017028093.1 | 20 | 3080 | XP_016883582.1 | |
| XM_017028094.1 | 20 | 2584 | XP_016883583.1 | |
| XM_017028095.1 | 19 | 2941 | XP_016883584.1 | |
| XM_017028096.1 | 21 | 2961 | XP_016883585.1 | |
| AF336127.1 | 19 | 2704 | AAK16734.1 | |
| AK075303.1 | 14 | 1979 | BAC11536.1 | |
| AK091157.1 | 20 | 2706 | ||
| AK296508.1 | 20 | 2862 | ||
| AK296760.1 | 20 | 2661 | ||
| BC110540.1 | 19 | 2677 | AAI10541.1 | |
| BC110541.1 | 18 | 2761 | AAI10542.1 | |
| BM666654.1 | 1 | 403 | ||
| HY211104.1 | 1 | 388 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067207 | Chr.20:3191285 - 3404426 on Build GRCh38 | Gain |
|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv952979 | Chr.20:3219555 - 3244454 on Build GRCh38 | Deletion |
|
| nsv528355 | Chr.20:3190691 - 3242579 on Build GRCh38 | Loss |
|
| nsv833898 | Chr.20:3147423 - 3309891 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map