Assay Details
Target Gene Details
Entrez Gene ID: | 390 |
Gene Name: | Rho family GTPase 3 |
Gene Aliases: |
ARHE, Rho8, RhoE, memB |
Location: |
Chr.2:150468193-150487695 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND3 | NM_001254738.1 | 5 | 1130 | NP_001241667.1 |
| NM_005168.4 | 6 | 1037 | NP_005159.1 | |
| AK298007.1 | 5 | 656 | ||
| AK299731.1 | 5 | 813 | ||
| AK309442.1 | 4 | 711 | ||
| AK311312.1 | 2 | 604 | ||
| BC012513.1 | 6 | 992 | AAH12513.1 | |
| S82240.1 | AAB47133.1 | |||
| X97758.1 | 5 | 1074 | CAA66352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834414 | Chr.2:150352860 - 150556795 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map