Assay Details
Target Gene Details
Entrez Gene ID: | 284525 |
Gene Name: | solute carrier family 9 member C2 (putative) |
Gene Aliases: |
SLC9A11 |
Location: |
Chr.1:173500465-173603094 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9C2 | NM_178527.3 | NP_848622.2 | ||
| XM_011509426.1 | XP_011507728.1 | |||
| XM_017001065.1 | XP_016856554.1 | |||
| XM_017001066.1 | XP_016856555.1 | |||
| XM_017001067.1 | XP_016856556.1 | |||
| XM_017001068.1 | XP_016856557.1 | |||
| XM_017001069.1 | XP_016856558.1 | |||
| XM_017001070.1 | XP_016856559.1 | |||
| XM_017001071.1 | XP_016856560.1 | |||
| XM_017001072.1 | XP_016856561.1 | |||
| XM_017001073.1 | XP_016856562.1 | |||
| XM_017001074.1 | XP_016856563.1 | |||
| XM_017001075.1 | XP_016856564.1 | |||
| XM_017001076.1 | XP_016856565.1 | |||
| AK128104.1 | ||||
| BC042592.1 | AAH42592.1 | |||
| BX648119.1 |
More Information
Set Membership: |
|

Genomic Map