Assay Details
Target Gene Details
Entrez Gene ID: | 3454 |
Gene Name: | interferon alpha and beta receptor subunit 1 |
Gene Aliases: |
AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC |
Location: |
Chr.21:33324443-33360361 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNAR1 | NM_000629.2 | NP_000620.2 | ||
| XM_005260964.2 | XP_005261021.1 | |||
| XM_011529552.1 | XP_011527854.1 | |||
| AK222770.1 | BAD96490.1 | |||
| AK222812.1 | BAD96532.1 | |||
| AK298051.1 | ||||
| AK312631.1 | ||||
| BC002590.2 | AAH02590.1 | |||
| BC021825.1 | AAH21825.1 | |||
| BG548299.1 | ||||
| J03171.1 | AAA52730.1 | |||
| JN543953.1 | ||||
| JN543954.1 | ||||
| JN543955.1 | ||||
| JN543956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587387 | Chr.21:33321963 - 33326080 on Build GRCh38 | Gain |
|
| nsv587394 | Chr.21:33323618 - 33325209 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map