Assay Details
Target Gene Details
Entrez Gene ID: | 953 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 1 |
Gene Aliases: |
ATPDase, CD39, NTPDase-1, SPG64 |
Location: |
Chr.10:95710901-95877266 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Target Gene Details
Entrez Gene ID: | 728558 |
Gene Name: | ENTPD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:95753206-96090235 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD1-AS1 | NR_038444.1 | 6 | 1207 | |
| AK091396.1 | 6 | 1186 |
More Information
Set Membership: |
|

Genomic Map