Assay Details
Target Gene Details
Entrez Gene ID: | 10278 |
Gene Name: | embryonal Fyn-associated substrate |
Gene Aliases: |
CAS3, CASS3, EFS1, EFS2, HEFS, SIN |
Location: |
Chr.14:23356400-23365633 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFS | NM_001277174.1 | 6 | 1923 | NP_001264103.1 |
| NM_005864.3 | 6 | 2430 | NP_005855.1 | |
| NM_032459.2 | 5 | 2151 | NP_115835.1 | |
| XM_005267256.1 | 5 | 1921 | XP_005267313.1 | |
| AB001466.1 | 6 | 2430 | BAA24588.1 | |
| AB001467.1 | 5 | 2151 | BAA24589.1 | |
| AK295933.1 | 6 | 1501 | ||
| BC034246.1 | 5 | 1699 | AAH34246.1 |
More Information
Set Membership: |
|

Genomic Map