Assay Details
Target Gene Details
Entrez Gene ID: | 10367 |
Gene Name: | mitochondrial calcium uptake 1 |
Gene Aliases: |
CALC, CBARA1, EFHA3, MPXPS |
Location: |
Chr.10:72367326-72626191 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICU1 | NM_001195518.1 | 12 | 2236 | NP_001182447.1 |
| NM_001195519.1 | 7 | 1586 | NP_001182448.1 | |
| NM_006077.3 | 13 | 2242 | NP_006068.2 | |
| XM_005269383.2 | 12 | 2158 | XP_005269440.1 | |
| XM_005269386.2 | 8 | 1699 | XP_005269443.1 | |
| XM_011539119.1 | 13 | 2308 | XP_011537421.1 | |
| AK022697.1 | 10 | 1701 | BAB14187.1 | |
| AK225298.1 | 12 | 2140 | ||
| AL117423.1 | 13 | 2133 | CAB55915.1 | |
| BC004190.2 | 12 | 2148 | AAH04190.1 | |
| BC004216.2 | 12 | 2148 | AAH04216.1 | |
| BC016641.1 | 12 | 2111 | AAH16641.1 | |
| BU627477.1 | 1 | 265 | ||
| Y17711.1 | 8 | 1564 | CAA76830.1 |
More Information
Set Membership: |
|

Genomic Map