Assay Details
Target Gene Details
Entrez Gene ID: | 59084 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative) |
Gene Aliases: |
NPP-5, NPP5 |
Location: |
Chr.6:46159182-46171045 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP5 | NM_001290072.1 | NP_001277001.1 | ||
| NM_001290073.1 | NP_001277002.1 | |||
| NM_021572.5 | NP_067547.1 | |||
| XM_005249260.3 | XP_005249317.1 | |||
| XM_011514786.2 | XP_011513088.1 | |||
| AK292842.1 | ||||
| AK295192.1 | ||||
| AK312790.1 | ||||
| AY358514.1 | AAQ88878.1 | |||
| BC027615.2 | AAH27615.1 | |||
| BX647968.1 | ||||
| DC321540.1 | ||||
| DC332590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576147 | Chr.6:46106919 - 46185586 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map