Assay Details
Target Gene Details
Entrez Gene ID: | 57579 |
Gene Name: | family with sequence similarity 135 member A |
Gene Aliases: |
KIAA1411 |
Location: |
Chr.6:70413404-70561174 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM135A | NM_001105531.2 | 1 | 234 | NP_001099001.1 |
| NM_020819.4 | 1 | 234 | NP_065870.3 | |
| XM_005248743.1 | 1 | 152 | XP_005248800.1 | |
| XM_005248744.4 | 1 | 205 | XP_005248801.1 | |
| XM_005248745.2 | 1 | 152 | XP_005248802.1 | |
| XM_005248748.2 | 1 | 152 | XP_005248805.1 | |
| XM_005248749.1 | 1 | 152 | XP_005248806.1 | |
| XM_005248751.2 | 1 | 152 | XP_005248808.1 | |
| XM_011535994.2 | 1 | 204 | XP_011534296.1 | |
| XM_011535995.2 | XP_011534297.1 | |||
| XM_011535996.2 | 1 | 204 | XP_011534298.1 | |
| XM_011535997.2 | 1 | 204 | XP_011534299.1 | |
| XM_017011124.1 | 1 | 219 | XP_016866613.1 | |
| XM_017011125.1 | XP_016866614.1 | |||
| XM_017011126.1 | 1 | 201 | XP_016866615.1 | |
| XM_017011127.1 | 1 | 165 | XP_016866616.1 | |
| XM_017011128.1 | 1 | 205 | XP_016866617.1 | |
| XM_017011129.1 | 1 | 220 | XP_016866618.1 | |
| XM_017011130.1 | 1 | 119 | XP_016866619.1 | |
| XM_017011131.1 | 1 | 88 | XP_016866620.1 | |
| XM_017011132.1 | 1 | 215 | XP_016866621.1 | |
| XM_017011133.1 | 1 | 121 | XP_016866622.1 | |
| XM_017011134.1 | 1 | 152 | XP_016866623.1 | |
| XM_017011135.1 | 1 | 206 | XP_016866624.1 | |
| BC131782.1 | ||||
| CR749254.1 | CAH18110.1 | |||
| DB462733.2 | 1 | 234 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524328 | Chr.6:70252130 - 70849102 on Build GRCh38 | Gain |
|
| nsv1027161 | Chr.6:70396954 - 70435463 on Build GRCh38 | Loss |
|
| nsv1027049 | Chr.6:70251907 - 70851785 on Build GRCh38 | Gain |
|
| nsv1032876 | Chr.6:70338725 - 70435463 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map