Assay Details
Target Gene Details
Entrez Gene ID: | 1052 |
Gene Name: | CCAAT/enhancer binding protein delta |
Gene Aliases: |
C/EBP-delta, CELF, CRP3, NF-IL6-beta |
Location: |
Chr.8:47736914-47738164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPD | NM_005195.3 | 1 | 300 | NP_005186.2 |
| BC094715.1 | AAH94715.1 | |||
| BC105109.1 | 1 | 295 | AAI05110.1 | |
| M83667.1 | 1 | 344 | AAA59927.1 |
More Information
Set Membership: |
|

Genomic Map