Assay Details
Target Gene Details
Entrez Gene ID: | 84342 |
Gene Name: | component of oligomeric golgi complex 8 |
Gene Aliases: |
CDG2H, DOR1 |
Location: |
Chr.16:69328621-69339623 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG8 | NM_032382.4 | NP_115758.3 | ||
| AK025968.1 | BAB15301.1 | |||
| AK056344.1 | BAB71157.1 | |||
| AK225391.1 | ||||
| AK302605.1 | ||||
| BC017492.1 | AAH17492.1 | |||
| BC121022.1 | ||||
| BC121023.1 |
Target Gene Details
Entrez Gene ID: | 64146 |
Gene Name: | peptide deformylase (mitochondrial) |
Gene Aliases: |
- |
Location: |
Chr.16:69328621-69330595 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NM_022341.1 | 1 | 197 | NP_071736.1 | |
| AF239156.1 | 1 | 197 | AAG33968.1 | |
| AF322879.1 | 1 | 172 | AAK15624.1 | |
| BC019912.1 | 1 | 173 | AAH19912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827715 | Chr.16:69311943 - 69340458 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map