Assay Details
Target Gene Details
Entrez Gene ID: | 3651 |
Gene Name: | pancreatic and duodenal homeobox 1 |
Gene Aliases: |
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1 |
Location: |
Chr.13:27919982-27926314 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDX1 | NM_000209.3 | 1 | 126 | NP_000200.1 |
| AF049893.1 | AAC05157.1 | |||
| CA771381.1 | 1 | 137 | ||
| CA848737.1 | 1 | 126 | ||
| U30329.1 | AAA74012.1 | |||
| U35632.1 | AAA88820.1 | |||
| X99894.1 | 1 | 117 | CAA68169.1 |
Target Gene Details
Entrez Gene ID: | 100861550 |
Gene Name: | PDX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:27829759-27921404 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDX1-AS1 | NR_047484.1 |
More Information
Set Membership: |
|

Genomic Map