Assay Details
Target Gene Details
Entrez Gene ID: | 55754 |
Gene Name: | transmembrane protein 30A |
Gene Aliases: |
C6orf67, CDC50A |
Location: |
Chr.6:75252922-75284916 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM30A | NM_001143958.1 | 6 | 2158 | NP_001137430.1 |
| NM_018247.3 | 7 | 2266 | NP_060717.1 | |
| AJ420517.1 | 1 | 939 | ||
| AK001718.1 | 7 | 2135 | BAA91859.1 | |
| AL162046.1 | 4 | 1494 | CAB82389.1 | |
| AL832490.1 | 7 | 2189 | CAH56205.1 | |
| AL832815.1 | 7 | 2128 | CAH56262.1 | |
| BC009006.1 | 6 | 2009 | AAH09006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603763 | Chr.6:75213889 - 75299710 on Build GRCh38 | Loss |
|
| nsv603762 | Chr.6:75079810 - 75270703 on Build GRCh38 | Loss |
|
| nsv603764 | Chr.6:75254035 - 75315887 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map