Assay Details
Target Gene Details
Entrez Gene ID: | 51263 |
Gene Name: | mitochondrial ribosomal protein L30 |
Gene Aliases: |
L28MT, L30MT, MRP-L28, MRP-L30, MRPL28, MRPL28M, RPML28 |
Location: |
Chr.2:99181079-99199557 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL30 | NM_145212.3 | 1 | 84 | NP_660213.1 |
| NR_028356.1 | 1 | 84 | ||
| AK025547.1 | ||||
| AK225894.1 | ||||
| BC000217.1 | ||||
| BC022391.1 | AAH22391.1 | |||
| BG532984.1 | ||||
| BU533068.1 | ||||
| DA310406.1 | 1 | 84 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3591732 | Chr.2:99123694 - 99182166 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map