Assay Details
Target Gene Details
Entrez Gene ID: | 10717 |
Gene Name: | adaptor related protein complex 4 beta 1 subunit |
Gene Aliases: |
BETA-4, CPSQ5, SPG47 |
Location: |
Chr.1:113894194-113905202 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4B1 | NM_001253852.2 | 8 | 1668 | NP_001240781.1 |
| NM_001253853.2 | 9 | 1593 | NP_001240782.1 | |
| NM_001308312.1 | 5 | 1164 | NP_001295241.1 | |
| NM_006594.4 | 9 | 1721 | NP_006585.2 | |
| XM_011540523.2 | 8 | 1574 | XP_011538825.1 | |
| XM_011540524.2 | 7 | 1408 | XP_011538826.1 | |
| XM_011540525.2 | 7 | 1520 | XP_011538827.1 | |
| XM_011540528.2 | 8 | 4426 | XP_011538830.1 | |
| XM_017000088.1 | 8 | 3405 | XP_016855577.1 | |
| XM_017000089.1 | XP_016855578.1 | |||
| XM_017000090.1 | 6 | 1159 | XP_016855579.1 | |
| XM_017000091.1 | XP_016855580.1 | |||
| XM_017000092.1 | 9 | 1785 | XP_016855581.1 | |
| AB209817.1 | 8 | 1605 | BAD93054.1 | |
| AF092094.1 | 9 | 1665 | AAD20448.1 | |
| AK001045.1 | 4 | 690 | ||
| AK057567.1 | 1 | 1634 | ||
| AK093740.1 | 5 | 1069 | ||
| AK300204.1 | 9 | 1432 | ||
| AK314641.1 | 9 | 1625 | ||
| BC014146.2 | 8 | 1526 | AAH14146.1 |
Target Gene Details
Entrez Gene ID: | 100287722 |
Gene Name: | AP4B1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:113812612-113901237 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4B1-AS1 | NR_037864.1 | |||
| NR_125965.1 | ||||
| AK123199.1 | ||||
| DB028504.1 | ||||
| DB074478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587251 | Chr.1:113871703 - 113936287 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map