Assay Details
Target Gene Details
Entrez Gene ID: | 56954 |
Gene Name: | nitrilase family member 2 |
Gene Aliases: |
HEL-S-8a |
Location: |
Chr.3:100334718-100355634 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIT2 | NM_020202.4 | 3 | 268 | NP_064587.1 |
| AF260334.1 | 3 | 267 | AAG44665.1 | |
| AF284574.1 | 3 | 215 | AAF87103.1 | |
| AK225979.1 | 3 | 244 | ||
| AK294010.1 | 3 | 229 | ||
| AK295835.1 | 3 | 229 | ||
| AK313704.1 | 3 | 210 | ||
| BC020620.1 | 3 | 225 | AAH20620.1 | |
| BC107890.1 | 3 | 256 | AAI07891.1 | |
| DB466042.1 | 3 | 268 | ||
| FJ224326.1 | 3 | 215 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3387925 | Chr.3:100338518 - 100340016 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map