Assay Details
Target Gene Details
Entrez Gene ID: | 55728 |
Gene Name: | NEDD4 binding protein 2 |
Gene Aliases: |
B3BP |
Location: |
Chr.4:40056223-40187039 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4BP2 | NM_001318359.1 | NP_001305288.1 | ||
| NM_018177.5 | NP_060647.2 | |||
| XM_006714023.3 | XP_006714086.1 | |||
| XM_011513716.2 | XP_011512018.1 | |||
| XM_011513717.2 | XP_011512019.1 | |||
| XM_011513718.2 | XP_011512020.1 | |||
| XM_011513719.2 | XP_011512021.1 | |||
| XM_011513720.1 | XP_011512022.1 | |||
| XM_011513721.1 | XP_011512023.1 | |||
| XM_017008397.1 | XP_016863886.1 | |||
| XM_017008398.1 | XP_016863887.1 | |||
| XM_017008399.1 | XP_016863888.1 | |||
| AB371584.1 | ||||
| AK001542.1 | BAA91748.1 | |||
| AU129951.1 | ||||
| AY267013.1 | AAP22172.1 | |||
| BC126466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010235 | Chr.4:39916983 - 40186134 on Build GRCh38 | Gain |
|
| nsv461341 | Chr.4:39983318 - 40185488 on Build GRCh38 | Gain |
|
| nsv594032 | Chr.4:39893368 - 40131994 on Build GRCh38 | Gain |
|
| nsv594050 | Chr.4:40073122 - 40167804 on Build GRCh38 | Loss |
|
| esv3575688 | Chr.4:39922679 - 40113190 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map