Assay Details
Target Gene Details
Entrez Gene ID: | 2896 |
Gene Name: | granulin |
Gene Aliases: |
CLN11, GEP, GP88, PCDGF, PEPI, PGRN |
Location: |
Chr.17:44345086-44353106 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRN | NM_002087.3 | NP_002078.1 | ||
| XM_005257253.1 | XP_005257310.1 | |||
| AF055008.1 | AAC09359.1 | |||
| AK000607.1 | ||||
| AK023348.1 | BAB14535.1 | |||
| AK222522.1 | BAD96242.1 | |||
| AK296090.1 | ||||
| AK303830.1 | ||||
| AY124489.1 | AAM94026.1 | |||
| BC000324.2 | AAH00324.1 | |||
| BC010577.2 | AAH10577.1 | |||
| BT006844.1 | AAP35490.1 | |||
| M75161.1 | AAA58617.1 | |||
| X62320.1 | CAA44196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5606n54 | Chr.17:44350262 - 44355375 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map