Assay Details
Target Gene Details
Entrez Gene ID: | 55779 |
Gene Name: | cilia and flagella associated protein 44 |
Gene Aliases: |
WDR52 |
Location: |
Chr.3:113286930-113445190 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP44 | XM_006713696.2 | XP_006713759.1 | ||
| XM_011512977.2 | XP_011511279.1 | |||
| XM_011512980.2 | XP_011511282.1 | |||
| XM_011512981.1 | XP_011511283.1 | |||
| XM_011512982.2 | XP_011511284.1 | |||
| XM_011512983.2 | XP_011511285.1 | |||
| XM_011512984.2 | XP_011511286.1 | |||
| XM_017006842.1 | XP_016862331.1 | |||
| XM_017006843.1 | XP_016862332.1 | |||
| XM_017006844.1 | XP_016862333.1 | |||
| XM_017006845.1 | XP_016862334.1 | |||
| XM_017006848.1 | XP_016862337.1 | |||
| XM_017006849.1 | XP_016862338.1 |
Target Gene Details
Entrez Gene ID: | 152185 |
Gene Name: | spindle and centriole associated protein 1 |
Gene Aliases: |
CCDC52, SPICE |
Location: |
Chr.3:113442718-113515187 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPICE1 | NM_144718.3 | 18 | 4199 | NP_653319.1 |
| AL832148.1 | 18 | 4168 | CAD91167.1 | |
| U79249.1 | 1 | 191 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map