Assay Details
Target Gene Details
Entrez Gene ID: | 22879 |
Gene Name: | MON1 homolog B, secretory trafficking associated |
Gene Aliases: |
HSRG1, SAND2, SRG1 |
Location: |
Chr.16:77190919-77199646 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MON1B | NM_001286639.1 | 5 | 2875 | NP_001273568.1 |
| NM_001286640.1 | 4 | 2717 | NP_001273569.1 | |
| NM_014940.3 | 6 | 3202 | NP_055755.1 | |
| AB020679.1 | 6 | 2886 | ||
| AK023374.1 | ||||
| AK128411.1 | 3 | 2744 | BAC87427.1 | |
| BU682558.1 | 1 | 147 |
Target Gene Details
Entrez Gene ID: | 100130958 |
Gene Name: | synaptonemal complex central element protein 1 like |
Gene Aliases: |
MRP2 |
Location: |
Chr.16:77199452-77213079 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYCE1L | NM_001129979.1 | NP_001123451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056676 | Chr.16:77105785 - 77820951 on Build GRCh38 | Loss |
|
| nsv1055234 | Chr.16:76969529 - 77433389 on Build GRCh38 | Gain |
|
| dgv3027n100 | Chr.16:76614676 - 77688151 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112674196] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map